Du sökte efter: Elektroforessystem
100  results were found
SearchResultCount:"100"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Artikel nummer : (700-0741)
Leverantör: VWR Collection
Beskrivning: The smallest of all the PerfectBlue electrophoresis systems, the Mini S can be used for up to 24 samples enabling short and rapid sample separation. T...
UOM: 1 * 1 Items
Leverantör: VWR Collection
Beskrivning: For 16×14 cm gels, the Twin M is a versatile system for SDS PAGE applications, in addition to non denaturing electrophoresis of proteins, SSCPs and di...
Leverantör: VWR Collection
Beskrivning: The largest of the series, the Twin L can accommodate 20×20 cm gels for the simultaneous separation of up to 50 samples making this system ideally sui...
Leverantör: VWR Collection
Beskrivning: Designed for ease of use and safe operation, the shiroGEL horizontal electrophoresis systems and accessories are built to withstand the rigours of eve...
Artikel nummer : (700-0292)
Leverantör: VWR Collection
Beskrivning: The shiroGEL mini vertical PAGE system is modular, allowing PAGE and electroblotting to be carried out in the same tank, simply by changing the insert...
UOM: 1 * 1 Items
Leverantör: VWR Collection
Beskrivning: The PerfectBlue Twin S system is designed for rapid separation of typical 10×10 cm mini gels. Due to its small size and a separation capacity of 6 to ...
Leverantör: VWR Collection
Beskrivning: The shiroGEL vertical 20 offers application flexibility and is distinguished by its ease of gel casting and set-up, requiring only four screws to secu...
Artikel nummer : (700-0910)
Leverantör: VWR Collection
Beskrivning: This 'extra wide' Maxi tank was developed especially for large sample numbers and rapid separation times. Accommodating up to 10 microtitre combs, the...
UOM: 1 * 1 Items
Leverantör: VWR Collection
Beskrivning: VWR® Real Time electrophoresis systems have revolutionised DNA electrophoresis. it is no longer necessary to remove a gel from a tank, place it onto a...
Artikel nummer : (700-0913)
Leverantör: VWR Collection
Beskrivning: The Maxi L system can be used to run up to 600 samples, making it perfectly suited for large scale sample screening where up to 12 microtitre combs ca...
UOM: 1 * 1 Items
Leverantör: VWR Collection
Beskrivning: Extra wide, for gels 20×10 cm, Twin ExW S is ideally suited to rapid analytical separations or large sample separation. The combination of the triple-...
Leverantör: VWR Collection
Beskrivning: The JustCast system is a flexible and quick system for casting multiple PerfectBlue™ compatible 'mini' gels at the same time, without the need to tape...
Artikel nummer : (700-0842)
Leverantör: VWR Collection
Beskrivning: With its extra wide design, the Midi ExW is ideal when there is a need to run lots of samples (up to 200) in just a short distance, for example, when ...
UOM: 1 * 1 Items
Artikel nummer : (700-0003)
Leverantör: VWR Collection
Beskrivning: Mini Gel II är ett komplett elektroforessystem som omfattar alla komponenter som krävs för att gjuta och köra horisontella geler - gelbrickor, kammar,...
UOM: 1 * 1 Items
Artikel nummer : (700-0912)
Leverantör: VWR Collection
Beskrivning: The Maxi ExW 'Revolution' offers built-in buffer recirculation - without the need for pumps or additional tubing. The Maxi ExW 'Revolution' offers the...
UOM: 1 * 1 Items
Artikel nummer : (730-0340)
Leverantör: VWR Collection
Beskrivning: This 96-well format horizontal gel electrophoresis unit is compatible with 8-channel multichannel pipettes, and matches up with a standard 96-well pla...
UOM: 1 * 1 Items
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 100
no targeter for Bottom