Du sökte efter: Diskmaskin för glasvaror
Diskmaskiner från leverantörer som Miele, Smeg och Labconco. Diskmaskinerna är anpassade för laboratoriemiljö. Olika korgar och insatser väljs till för att anpassa dina behov.
Diskmaskiner från leverantörer som Miele, Smeg och Labconco. Diskmaskinerna är anpassade för laboratoriemiljö. Olika korgar och insatser väljs till för att anpassa dina behov.
17  results were found
SearchResultCount:"17"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Leverantör: MIELE
Beskrivning: The PLW 8616 laboratory washer can accommodate 216 laboratory flasks, 588 vials, or 294 pipettes. The large glassware washer is flexible and simple to...
Leverantör: MIELE
Beskrivning: Miele PG 8583 Laboratory glasswasher represents a professional solution for laboratory glassware for analytical experiments. This approach is particul...
Leverantör: MIELE
Beskrivning: Miele PG 8593 laboratory glasswasher represents a professional solution for laboratory glassware for analytical experiments. This approach is particul...
Leverantör: MIELE
Beskrivning: The PLW 6111 SlimLine laboratory washer offer a comprehensive and systematic solution to the reprocessing of laboratory glassware for analytical exper...
Leverantör: MIELE
Beskrivning: Miele PG 8583 CD Laboratory glasswasher represents a professional solution for laboratory glassware for analytical experiments. This approach is parti...
Leverantör: MIELE
Beskrivning: The PLW 8615 large laboratory glasswasher can accommodate 216 laboratory flasks, 588 vials, or 294 pipettes. The large glasswasher is flexible and sim...
Leverantör: MIELE
Beskrivning: The PLW 8617 large laboratory glasswasher can accommodate 216 laboratory flasks, 588 vials, or 294 pipettes. The large glasswasher is flexible and sim...
Leverantör: MIELE
Beskrivning: Glassware washing machines, APS 102 Sego, SegoAssign Plus To connect the Segosoft with the in-house patient management system, Ø 2,5×80 mm, B×D×H (mm): Ø 2.5×80 mm
Leverantör: MIELE
Beskrivning: PG 8504 is a freestanding unit with lid and its exterior is made from stainless steel casing. It can handle a wide variety of cleaning challenges foun...
Leverantör: MIELE
Beskrivning: The PLW 7111 SlimLine laboratory washers offer a comprehensive and systematic solution for analytically pure reprocessing of laboratory glassware.
Leverantör: MIELE
Beskrivning: G 7896 DOS cabinet: for 1 to 4 DOS modules with containers.
Leverantör: SMEG S.P.A.
Beskrivning: This range of professional Smeg washing and thermal disinfection machines are manufactured using high quality materials and components in order to ach...
Artikel nummer : (SMEGGW4090P)
Leverantör: SMEG S.P.A.
Beskrivning: Display: 128×64 pixel.
UOM: 1 * 1 Items
New Product
Leverantör: SMEG S.P.A.
Beskrivning: The range of professional Smeg washing and disinfection machines is manufactured according to specific projects using professional materials and speci...
Artikel nummer : (BELAF389600000)
Leverantör: Bel-Art Products, a Part of SP
Beskrivning: Economical, durable and gentle all-plastic washer for delicate glass or quartz cuvettes.
UOM: 1 * 1 Items
Artikel nummer : (141-0012)
Leverantör: GETINGE LIFE SCIENCES
Beskrivning: The Lancer 815 LX undercounter glassware washer fits neatly in the standard 60 cm laboratory casework, offers professional features and the same high ...
UOM: 1 * 1 Items
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 17
no targeter for Bottom