Anaerobic systems, workstation, Whitley DG250
Leverantör: Don Whitley Scientific
390-1980EA 0 SEK
DOWIA03000
Anaerobic systems, workstation, Whitley DG250
The DG250 is an entry-level workstation, providing you with a generous working area and large incubation capacity in the smallest possible footprint.
  • Perfect for laboratories growing anaerobes and microaerophiles where space is limited - only 810 mm of bench width required
  • Each user porthole acts as a mini-airlock so you can transfer plates at the same time as entering/exiting the workstation
  • The complete interior is temperature controlled – and can be set between 5 °C above ambient and 45 °C
  • The Whitley DG250 has an impressive 33% more capacity than any other unit of comparable size, so you get more for your money
  • The whole top can be lifted off for thorough cleaning, for bulk sample transfer and to introduce/remove equipment
  • Automatic dehumidifier – no user maintenance necessary
For emergency anaerobic incubation it will hold up to 400 x 90mm Petri dishes. This workstation can be used for both anaerobic and microaerobic applications simply by changing the gas supplies.
Note: The cycle time mentioned in the specifications is for the airlock.
Beställningsinformation: Options and accessories not included (sold separately). For more information on the available options and accessories, please see the attached brochures and contact your Avantor representative.