Du sökte efter: Fatpumpar
94  results were found
SearchResultCount:"94"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Leverantör: Merck
Beskrivning: Merck offers a broad variety of withdrawal systems that are tailor-made for barrels and drums. They are designed to ensure safe, easy and efficient so...
Artikel nummer : (MFLX07571-02)
Leverantör: Avantor Fluid Handling
Beskrivning: Sample depths down to 7.9 meters (26 feet).
UOM: 1 * 1 Items
CE Compliant CSA Certified UL Listed
Artikel nummer : (99901.010)
Leverantör: VWR Chemicals
Beskrivning: Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 10 L stainless steel barrels.
UOM: 1 * 1 Items
Artikel nummer : (99900.001)
Leverantör: VWR Chemicals
Beskrivning: Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 10 and 30 L stainless steel barrels.
UOM: 1 * 1 Items
Artikel nummer : (99907.030)
Leverantör: VWR Chemicals
Beskrivning: Specially developed solvent withdrawal system for inert gas pressurizing.
UOM: 1 * 1 Items
Artikel nummer : (99901.030)
Leverantör: VWR Chemicals
Beskrivning: Specially developed solvent withdrawal system with manual pressure build-up for VWR® returnable 30 L stainless steel barrels.
UOM: 1 * 1 Items
Artikel nummer : (99907.010)
Leverantör: VWR Chemicals
Beskrivning: Specially developed solvent withdrawal system for inert gas pressurizing.
UOM: 1 * 1 Items
Leverantör: BÜRKLE
Beskrivning: Barrel pump, made of stainless steel AISI 304 (1.4301), suitable for filling combustible and easily flammable liquids. It can conduct electricity and,...
Leverantör: BÜRKLE
Beskrivning: Idealisk för avtappning av vätskor med hög renhet. PTFE och FEP har hög kemikalieresistens vilket möjliggör pumpning av vätskor som påverkar vanliga m...
Leverantör: BÜRKLE
Beskrivning: Drum pump, Fatpump med slang och kran, Material: PP, Flödeshastighet: 200 ml/slag, Nedsänkning: 500 mm, Längd: 600 mm
Leverantör: KNF
Beskrivning: Simple, user-friendly compact dosing pump.
Application: Metering liquids.
Leverantör: BÜRKLE
Beskrivning: A bomba alimentada por unidade de alimentação elétrica, EnergyOne, possui um novo sistema híbrido exclusivo completamente flexível e independente do c...
Leverantör: KNF
Beskrivning: These diaphragm liquid pumps work quietly and with little vibration to pump neutral and aggressive liquids. Application: Transferring liquids.
Artikel nummer : (300-0359)
Leverantör: BÜRKLE
Beskrivning: OTAL® pump with disposable delivery tube made of PE. In OTAL® pumps the medium being transferred only comes in contact with the delivery tube. The dis...
UOM: 1 * 1 Items
Artikel nummer : (300-0360)
Leverantör: BÜRKLE
Beskrivning: OTAL® pump with disposable delivery tube made of PE. In OTAL® pumps the medium being transferred only comes in contact with the delivery tube. The dis...
UOM: 1 * 1 Items
Artikel nummer : (223-0123)
Leverantör: BÜRKLE
Beskrivning: In the withdrawal system for solvents with discharge hose, the barrel pump has a flexible discharge hose. The hose makes it possible to dispense the r...
UOM: 1 * 1 Items
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 94
no targeter for Bottom