Du sökte efter: Sterilisatorer
Sterilisatorer för torrsterilisering av kirurgiska instrument,laboratorieinstrument och andra material. Sterilisatorerna finns som fristående eller bänkversioner.
Sterilisatorer för torrsterilisering av kirurgiska instrument,laboratorieinstrument och andra material. Sterilisatorerna finns som fristående eller bänkversioner.
32  results were found
SearchResultCount:"32"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Leverantör: VWR Collection
Beskrivning: Easy to use, automatic, flexible, reliable and convenient autoclave for all standard laboratory procedures with special features, such as F₀ calculati...
Leverantör: VWR Collection
Beskrivning: These autoclaves are ideal for media, equipment and waste sterilisation. Designed for safety level laboratories, they are equipped with exhaust air fi...
Sale
Leverantör: VWR Collection
Beskrivning: VAPOUR-Line autoclaves are designed for applications such as liquids in open or slightly closed vessels, instruments and solids, unwrapped and waste s...
Leverantör: VWR Collection
Beskrivning: Autoclave for occasional use. AISI 304 stainless steel housing, easy to maintain and disinfect, aluminium inner chamber and ABS insulating lid with lo...
Sale
Leverantör: TUTTNAUER
Beskrivning: Tuttnauer's laboratory autoclaves are ideal for a wide range of applications in various fields, such as research institutes, universities, pharmaceuti...
Leverantör: MEMMERT
Beskrivning: This steriliser complies with all relevant standards and regulations for medical products and guarantees reliable programme sequences, irrespective of...
Leverantör: TUTTNAUER
Beskrivning: Tuttnauer's laboratory autoclaves are ideal for a wide range of applications in various fields, such as research institutes, universities, pharmaceuti...
Leverantör: MEMMERT
Beskrivning: This steriliser complies with all relevant standards and regulations for medical products and guarantees reliable programme sequences, irrespective of...
Leverantör: MEMMERT
Beskrivning: These sterilisers comply with all relevant standards and regulations for medical products, and guarantee reliable program sequences, irrespective of t...
Artikel nummer : (TUTT3850EAP-D)
Leverantör: TUTTNAUER
Beskrivning: The EA autoclave significantly increase your productivity with extra fast and efficient drying cycles. These two models have the added benefit of a hi...
UOM: 1 * 1 Items
Leverantör: TUTTNAUER
Beskrivning: Affordable performance and trusted reliability for your laboratory sterilisation needs. These compact units sterilise liquids, solids, sealed and unse...
Leverantör: TUTTNAUER
Beskrivning: The Classic is reliable and trusted worldwide, designed to give you the most cost effective sterilisation solution. This light, compact, portable and ...
Leverantör: TUTTNAUER
Beskrivning: Tuttnauer's ML laboratory line of semi-automatic, electro-mechanically controlled, vertical steam sterilizers provide safe, economical and effective s...
Artikel nummer : (481-2004)
Leverantör: TUTTNAUER
Beskrivning: Horizontal and vertical autoclave with automatic control. The chamber is constructed of long lasting 316L/Ti grade stainless steel with superior corro...
UOM: 1 * 1 Items
Leverantör: Priorclave
Beskrivning: These versatile autoclaves are fully equipped for use in laboratories throughout the food, drink, dairy, health and pharmaceutical industries and offe...
Leverantör: BIOTOOL AG
Beskrivning: Electrical sterilisation systems which are suitable for sterilising loops, needles and instruments. Ideal for use in laboratories where using gases or...
Sale
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 32
no targeter for Bottom