Du sökte efter: Vakuumsugsystem
17  results were found
SearchResultCount:"17"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Leverantör: Integra
Beskrivning: Används för ren och professionell uppsugning av biologiska och kemiska vätskor. Alla funktioner som kan behövas är inbyggda i det kompakta systemet. S...
Leverantör: Integra
Beskrivning: VACUBOY handkontroll är idealisk för uppsugning av alla sorters vätskor från kyvetter, mikrotiterplattor, provrör, petriskålar, cellodlingsflaskor, ro...
Leverantör: Integra
Beskrivning: In the laboratory small volumes of liquid need be disposed of on a daily basis. This includes the removal of excess liquids from reaction tubes, aspir...
Leverantör: VACUUBRAND GMBH + CO
Beskrivning: The VacuuCenter BVC basic is designed for connection to an existing external vacuum source like a diaphragm pump or a vacuum network (e.g., VACUU·LAN®...
Energy Efficient
Leverantör: VACUUBRAND GMBH + CO
Beskrivning: The VacuuCenter BVC control serves for efficient and convenient aspiration of supernatants. The vacuum level - and thus the suction force - is precise...
Energy Efficient
Leverantör: VACUUBRAND GMBH + CO
Beskrivning: The VacuuCenter BVC professional serves for aspiration and disposal of biological fluids, providing outstanding levels of comfort and safety for worki...
Energy Efficient
Artikel nummer : (181-9930)
Leverantör: Thermo Fisher Scientific
Beskrivning: Attach the Nalgene™ aspirator vacuum pump directly to sink faucets for vacuum draw. It can draw 11,5 L air per minute at a water flow rate of 6,5 L pe...
UOM: 1 * 1 Items
Artikel nummer : (BELAF199170250)
Leverantör: Bel-Art Products, a Part of SP
Beskrivning: These system is used to aspirate tissue culture media, supernatants, and chemical wastes.
UOM: 1 * 1 Items
Leverantör: GILSON
Beskrivning: The Gilson® safe aspiration station is a stand-alone motorised vacuum system for quick and easy aspiration of biological liquid waste disposal. Liquid...
Artikel nummer : (216-1098)
Leverantör: GRANT INSTRUMENTS LTD.
Beskrivning: All in one system with integrated pump designed for routine aspiration of the supernatent alcohol/buffer from the walls of microtubes during DNA/RNA p...
UOM: 1 * 1 Items
Leverantör: HAEP LABOR CONSULT
Beskrivning: The simple vacuum safety aspiration system, together with the quick lock adapters, filter, ventilation insert and the silicone tubing forms a complete...
Leverantör: HAEP LABOR CONSULT
Beskrivning: Vacuum suction systems for connection to laboratory vacuum system without own suction pump.
Artikel nummer : (HAEP981323101)
Leverantör: HAEP LABOR CONSULT
Beskrivning: Automatic suction system with suction capacity for connection to laboratory vacuum system.
UOM: 1 * 1 Items
Artikel nummer : (HAEP981323201)
Leverantör: HAEP LABOR CONSULT
Beskrivning: Automatic safety vacuum system for extraction.
UOM: 1 * 1 Items
Artikel nummer : (DOWIA07950)
Leverantör: DON WHITLEY SCIENTIFIC LTD
Beskrivning: This AsPROvac is an all-in-one laboratory aspirator to ensure biological liquid waste can be aspirated and disposed of quickly and safely.
UOM: 1 * 1 Items
Leverantör: KOEHLER TECHNISCHE PRODUKTEN
Beskrivning: Vacuum aspiration system. A plastic supporting plate with carrying handles and attachments for holding the vacuum-proof suction flask make the system ...
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 17
no targeter for Bottom