Du sökte efter: Vattenreningssystem
60  results were found
SearchResultCount:"60"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Leverantör: VWR Collection
Beskrivning: This 'two in one' lab water system produces ultrapure (Type I) and RO (Type III) water from tap water. Quality of ultrapure water meets or exceeds AST...
Leverantör: VWR Collection
Beskrivning: This powerful and intuitive laboratory water purification system produces Type I ultrapure water from RO, distilled or deionised water. The product wa...
Leverantör: VWR Collection
Beskrivning: This compact 'plug and play' lab water system is an ideal choice for users who need up to 20 litres of ultrapure Type I water or RO water (Type III) p...
Sale
Leverantör: VWR Collection
Beskrivning: This powerful, intuitive laboratory water purification system provides superior and consistent ultrapure Type I and EDI-based Type II water from tap w...
Sale
Leverantör: VWR Collection
Beskrivning: This compact system produces EDI-based Type II water directly from tap water. Quality of Type II water meets or exceeds water quality as defined by AS...
Sale
Artikel nummer : (171-2708)
Leverantör: VWR Collection
Beskrivning: This demineraliser connects to a mains water tap and instantly delivers pure water with high resistivity. The exchange capacity of a cartridge is 300&...
UOM: 1 * 1 Items
Leverantör: Merck
Beskrivning: To deliver reliable, high quality scientific results, you require consistently high quality ultrapure water. The Milli-Q® EQ 7000 ultrapure water syst...
Energy Efficient
Sustainable Materials
Waste Reducing
Leverantör: Merck
Beskrivning: Elix® Essential delivers Type 2 water customised to fit a broad range of research areas. It is designed to be fed from potable water and is the ideal ...
Artikel nummer : (172-5109)
Leverantör: Merck
Beskrivning: Kompakt, lättanvänd Milli-DI®-vattenrenare har utformats speciellt för framställning av små mängder rent vatten, mellan 3 och 5 liter per dag. Vattnet...
UOM: 1 * 1 Items
Leverantör: Merck
Beskrivning: Reliable water purification for results reproducibility and lab productivity.
Leverantör: Merck
Beskrivning: Purifies up to 3000 L/day with integrated storage, distribution and remote monitoring.
Leverantör: Merck
Beskrivning: Super-Q® Plus System is a high-flow water system designed to provide the 'final polish' to water which has been pretreated by reverse osmosis, distill...
Leverantör: Merck
Beskrivning: Direct-Q® 3 har utvecklats för enkel och direkt användning. Ultrarent vatten av typ I passar för beredning av buffertar för vätskekromatografi och bio...
Leverantör: MIELE
Beskrivning: Aqua purifier for two full or partial demineralisation cartridges. The casing made from white steel or stainless steel.
Leverantör: Merck
Beskrivning: RiOs™ Essential systems are ideal for laboratories needing reliable, constant production of Reverse Osmosis, RO (Type 3) water from potable tap water....
Artikel nummer : (BIND8012-0759)
Leverantör: Binder
Beskrivning: This system is used to fully desalinate faucet water for BINDER climate chamber humidification systems. The ion exchanger system consists of a pressur...
UOM: 1 * 1 Items
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 60
no targeter for Bottom