Vill du logga in igen?
Silvernitrat 0.1 mol/l (0.1 N) vattenlösning volumetrisk lösning
Leverantör: REAGECON
Varning
Formel: AgNO₃
MW: 169,87 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9,III
https://se.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg
ORDER
Expandera alla / Komprimera alla
Förp. VWR art.nr Antal
500 ml REGEN20100H
Expandera alla / Komprimera alla
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal