2,2,2-trifluoretanol, Supelco®
Leverantör: Merck
|
Fara
|
M-Clarity™ quality level = MQ100
91683-25MLEA
853
SEK
91683-25ML
2,2,2-trifluoretanol, Supelco®
2,2,2-trifluoretanol
Formel:
C₂H₃F₃O MW: 100,04 g/mol Kokpunkt: 74,1 °C (1013 hPa) Smältpunkt: –43,5 °C Densitet: 1,39 g/cm³ (20 °C) Flampunkt: 33 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00004672 CAS nummer: 75-89-8 EINECS: 200-913-6 UN: 2929 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...