n-Hexatriacontane, Supelco®


M-Clarity™ quality level = MQ100

SUPL52919-250MGEA 975 SEK
SUPL52919-250MG
n-Hexatriacontane, Supelco®
n-Hexatriacontane
Formel: H₃C(CH₂)₃₄CH₃
MW: 506,98 g/mol
Kokpunkt: 497 °C (1013 hPa)
Smältpunkt: 75,5 °C
Densitet: 0,7803 g/cm³ (80 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00009413
CAS nummer: 630-06-8
EINECS: 211-127-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR