Håll mig inloggad?
Avslutas om 
Din inloggning har gått ut
Din session har gått ut. För din säkerhet har vi loggat ut dig.
Vill du logga in igen?

Organisk referensstandard, Kloretan 100 µg/ml i metanol

Leverantör: LGC Standards
Dr Ehrenstorfer



 

ORDER

Expandera alla / Komprimera alla
Benämning Förp. VWR art.nr Antal
Kloretan 100 µg/ml i metanol 1 ml EHERXA11409000ME
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFIKATIONER

Benämning Chloroethane
CAS-nr 75-00-3
Formula C₂H₅Cl
Enter Details

Select the article for which you are looking for Certificate

Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping