Vill du logga in igen?
Thiosalicylic acid
Leverantör: Molekula
Varning
Formel: C₇H₆O₂S
MW: 154,19 g/mol
Kokpunkt: 230 °C (0,02 mmHg)
Smältpunkt: 165…166 °C
Densitet: 1,489 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004836
CAS nummer: 147-93-3
EINECS: 205-704-3
UN: 3335
ADR: 9,
Merck Index: 12,09498
ORDER
Expandera alla / Komprimera alla
Förp. VWR art.nr Antal
100 g MOLEM63776863
Expandera alla / Komprimera alla
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal