3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT), Sigma-Aldrich®
Leverantör: Merck Millipore Calbiochem
|
Varning
|
475989-10EA
13875
SEK
475989-10
475989-1
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT), Sigma-Aldrich®
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Formel:
C₁₈H₁₆N₅BrS MW: 414,33 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00011964 CAS nummer: 298-93-1 EINECS: 206-069-5 |