Vill du logga in igen?
Spectrophotometers, GeneQuant™ 1300
Leverantör: Biochrom
GeneQuant™
The GeneQuant 1300 spectrophotometer is pre-programmed with methods for nucleic acid purification, bacterial cell cultures, protein assays, Cy™Dye and cell culture density measurement.
  • Other dyes into purified microarray hybridisation probes and PCR products
  • Zoom facility, peak identification and on-peak confirmation
  • Nucleic acid scanning to identify impurities, especially useful with RNA samples
cDNA application software for measuring incorporation of Cy™3 and Cy™5.
Leveransinformation: USB cable and print via computer software supplied as standard.
ORDER
Expandera alla / Komprimera alla
Benämning Modell Förp. VWR art.nr Antal
Spectrophotometer GeneQuant™ 1300 1 SCLI80-2120-00
Spectrophotometer with printer GeneQuant™ 1300 1 SCLI80-2120-01
Spectrophotometer with Bluetooth GeneQuant™ 1300 1 SCLI80-2120-02
Cells
Benämning Höjd Volym Skiktdjup Material Förp. VWR art.nr Antal
Quartz cell - 5 – 7 µl 5 mm Quartz 1 SCLI80-2103-68
Cell with stopper and black walls - 70 µl 10 mm Quartz 1 SCLI80-2103-69
Cell with stopper and black walls - 750 µl 10 mm Quartz 1 SCLI80-2002-77
Rectangular cell with lid 45 mm 2 ml 10 mm UV silica 1 SCLI80-2002-58
Capillary cell for Genequant 100/300 - - 0,5 mm Quartz 1 SCLI80-2120-19
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Display LCD
Ljuskälla Pulsed Xenon
Våglängdsområde (nm) 190 - 1100 ; 200 - 900 in less than 5 seconds
Våglängdsområde 190 − 1100 nm
TILLBEHÖR
Expandera alla / Komprimera alla
Accessories
Benämning Höjd Volym Skiktdjup Material Förp. VWR art.nr Antal
Spare capillaries - 3 µl 0,5 mm Quartz 100 SCLI80-2104-67
Quartz cell - 5 – 7 µl 5 mm Quartz 1 SCLI80-2103-68
Cell with stopper and black walls - 70 µl 10 mm Quartz 1 SCLI80-2103-69
Cell with stopper and black walls - 750 µl 10 mm Quartz 1 SCLI80-2002-77
Rectangular cell with lid 45 mm 2 ml 10 mm UV silica 1 SCLI80-2002-58
Capillary cell for Genequant 100/300 - - 0,5 mm Quartz 1 SCLI80-2120-19
Bluetooth device - - - - 1 SCLI28918225
Printer - - - - 1 SCLI28918227
Spare papers for printer - - - - 20 Roll SCLI28918226
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal