Vill du logga in igen?
Spectrophotometers, GeneQuant™ 100
Leverantör: Biochrom
GeneQuant™
GeneQuant 100 is a compact spectrophotometer with predefined methods for nucleic acid purification, protein assays and cell culture density measurements.
  • Built-in printer (optional) or data output via USB to a PC for printing or storage
  • Optics with no moving parts for high energy throughput combined with a xenon source for long lamp lifetime
  • Three year lamp warranty
Visualisation of nucleic acid scans allow impurities to be detected particularly useful for RNA samples.
Proteins can be quantified by BCA, Biuret, Bradford, Direct UV and Lowry methods.
ORDER
Expandera alla / Komprimera alla
Benämning Förp. VWR art.nr Antal
Spectrophotometer 1 SCLI80-2130-00
Spectrophotometer with printer 1 SCLI80-2130-01
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Display Graphical LCD
Optiskt system Double beam
Utgång USB
Ljuskälla Pulsed Xenon
Spektral bandbredd 5 nm
Våglängdsområde 190 − 900 nm
Våglängdsnoggrannhet ±2 nm
TILLBEHÖR
Expandera alla / Komprimera alla
Accessories
Benämning Volym Skiktdjup Material Förp. VWR art.nr Antal
Spare capillaries 3 µl 0,5 mm Quartz 100 SCLI80-2104-67
Quartz cell 5 – 7 µl 5 mm Quartz 1 SCLI80-2103-68
Cell with stopper and black walls 70 µl 10 mm Quartz 1 SCLI80-2103-69
Cell with stopper and black walls 750 µl 10 mm Quartz 1 SCLI80-2002-77
Capillary cell for Genequant 100/300 - 0,5 mm Quartz 1 SCLI80-2120-19
PVC software - - - 1 SCLI28923188
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal