Vill du logga in igen?
Cuvettes for UV/Vis spectrophometers
Leverantör: Biochrom
ORDER
Expandera alla / Komprimera alla
Benämning Volym Skiktdjup Material Förp. VWR art.nr Antal
Capillary cell 3 µl 0,5 mm Quartz 1 SCLI80-2104-66
Spare capillaries 3 µl 0,5 mm Quartz 100 SCLI80-2104-67
Quartz cell 5 – 7 µl 5 mm Quartz 1 SCLI80-2103-68
Cell with stopper and black walls 70 µl 10 mm Quartz 1 SCLI80-2103-69
Cell with lid and black walls 400 µl 10 mm Quartz 1 SCLI80-2002-95
Cell with stopper and black walls 750 µl 10 mm Quartz 1 SCLI80-2002-77
Cell with stopper and black walls 400 μl 10 mm Quartz 1 SCLI80-2002-99
Cell with stopper and black walls 800 µl 10 mm Quartz 1 SCLI80-2002-81
Capillary cell for Genequant 100/300 - 0,5 mm Quartz 1 SCLI80-2120-19
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal