Vill du logga in igen?
Drum pumps, AccuOne and EnergyOne
Leverantör: Bürkle
The barrel pumps AccuOne and EnergyOne with the unique hybrid system ensure fast, flexible and convenient filling. The pumps are equipped with two different power supply systems: AccuOne, driven by a high-performance rechargeable battery or EnergyOne, with a power cable for direct connection to the electrical network. AccuOne and EnergyOne are designed for applications in which even very small amounts of liquid can be added to containers with narrow openings. They are made of high quality materials PP, PTFE and Hastelloy C, to ensure outstanding chemical resistance. This also makes them suitable for dispensing acids and bases.
  • Unique hybrid system, rechargeable battery and power supply unit are interchangeable
  • The TouchPanel features a foil keyboard sealed off against dust, dirt and water
  • Excellent residual draining and pumping capacity
  • 1 drive for several pump stations
  • Outstanding chemical resistance
The rechargeable battery or the power supply unit are fitted with a clip lock and are easily removable. The AccuOne can also be operated with the power supply cable unit and EnergyOne can be operated with the rechargeable battery. With a replacement rechargeable battery as an accessory, the user never has to wait for recharging. The drive can also be disconnected quickly and easily from the pump station with a clip lock.
AccuOne
Lithium-ion rechargeable battery
Replacement back-up battery available
Up to 600 litres of delivery volume per battery charge
EnergyOne
Unlimited filling duration
Available with power supply unit or rechargeable battery (available as accessory)
With power supply unit and fixed discharge elbow
ORDER
Expandera alla / Komprimera alla
AccuOne
Benämning Material Flödeshastighet Nedsänkning Förp. VWR art.nr Antal
AccuOne barrel pump PP/Hastelloy C/PTFE 0,4 - 12 l/min 700 mm 1 300-0085
-
AccuOne barrel pump PP/Hastelloy C/PTFE 0,4 - 12 l/min 1000 mm 1 300-0086
-
AccuOne barrel pump PP/Hastelloy C/PTFE 0,4 - 12 l/min 500 mm 1 300-0084
-
EnergyOne
Benämning Material Flödeshastighet Nedsänkning Förp. VWR art.nr Antal
EnergyOne barrel pump PP/Hastelloy C/PTFE 0,4 - 12 l/min 700 mm 1 300-0088
-
EnergyOne barrel pump PP/Hastelloy C/PTFE 0,4 - 12 l/min 500 mm 1 300-0087
-
EnergyOne barrel pump PP/Hastelloy C/PTFE 0,4 - 12 l/min 1000 mm 1 300-0089
-
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
TILLBEHÖR
Expandera alla / Komprimera alla
Accessories
Benämning Förp. VWR art.nr Antal
Fatanslutning för Mauser 2", intern gänga 1 223-1262
-
Fatanslutning för R2", intern gänga 1 223-1261
-
Fatanslutning för Tri-Sure", intern gänga 1 223-1263
-
Batteriladdare för Accu/EnergyOne 1 300-0091
-
Enhet utan laddningsbart batteri/nätaggregat 1 300-0093
-
Laddningsbart batteri utan laddare för Accu/EnergyOne 1 300-0090
-
Pumpstation Accu/EnergyOne,
dränkbar, längd 100 mm
1 300-0098
-
Pumpstation Accu/EnergyOne,
dränkbar, längd 70 mm
1 300-0097
-
Pumpstation Accu/EnergyOne, dränkbar, längd 50 mm 1 300-0096
-
Strömförsörjningsenhet inklusive anslutning 1 300-0092
-
Utloppsrör 1 300-0094
-
Utloppsslang med munstycke 1 300-0095
-
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal