Vill du logga in igen?
Membranfilter, typ 123
Leverantör: Sartorius
ORDER
Expandera alla / Komprimera alla
Typ Porstorlek Diam. Förp. VWR art.nr Antal
123 1,2 µm 25 mm 100 512-4213
123 1,2 µm 47 mm 100 512-4313
123 1,2 µm 50 mm 100 512-4413
123 1,2 µm 100 mm 25 512-4513
123 1,2 µm 142 mm 25 512-4613
123 1,2 µm 142 mm 100 514-0902
123 1,2 µm 293 mm 25 512-4713
123 5 µm 25 mm 100 512-4211
123 5 µm 47 mm 100 514-0096
123 5 µm 142 mm 25 514-0903
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFIKATIONER
Typ 123
Porstorlek 1,2 µm 5 µm 1,2 µm 5 µm 1,2 µm
Tjocklek 140 µm
Vattenflödeshastighet 320 ml/min/cm² (1 bar) 570 ml/min/cm² (1 bar) 320 ml/min/cm² (1 bar) 570 ml/min/cm² (1 bar) 320 ml/min/cm² (1 bar)
Max. temperatur (°C) 180
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal