Zirkonium(IV)oxid, Patinal®
Leverantör: Merck
1.08614.0100EA
0
SEK
1.08614.0100
1.08614.1000
Zirkonium(IV)oxid, Patinal®
Zirconium(IV) oxide
Formel:
ZrO₂ MW: 123,22 g/mol Kokpunkt: 4300 °C (1013 hPa) Smältpunkt: 2680 °C Densitet: 5,75 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00011310 CAS nummer: 1314-23-4 EINECS: 215-227-2 Merck Index: 14,10180 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...