Du sökte efter: Rengöringskit och dynor
25  results were found
SearchResultCount:"25"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Artikel nummer : (611-2886)
Leverantör: VWR Collection
Beskrivning: Cleaning kit containing a puffer brush (lens cleaning brush), cotton cloth and pair of nitrile gloves.
UOM: 1 * 1 KIT
Leverantör: CONTEC
Beskrivning: Disposable Premira II Microfibre HighDuster covers reduce fibre shedding and provide effective dry and damp dusting for hard to reach areas. The micro...
Artikel nummer : (BRDY018556)
Leverantör: Brady
Beskrivning: This cleaning kit for printers includes: Instruction sheet, five cleaning cards soaked in IPA and the material safety data sheet.
UOM: 1 * 1 KIT
Artikel nummer : (CONTMGMP0001)
Leverantör: CONTEC
Beskrivning: These single use microfibre pads feature synthetic construction for the effective removal of both gross contamination and fine particles. Conveniently...
UOM: 1 * 140 Items
Leverantör: Cytiva (Formerly Pall Lab)
Beskrivning: Handy dispenser kits allow one-handed dispensing of cellulose absorbent pads.
Artikel nummer : (115-1752)
Leverantör: 3M
Beskrivning: These hand pads can be used in place of steel wool for conditioning metal surfaces and paint prep. Unlike fine steel wool, Scotch-Brite™ pads will not...
UOM: 1 * 20 Items
Artikel nummer : (MMMA07904)
Leverantör: 3M
Beskrivning: Rengöringsdynor, Scotch-Brite™, Handhållare, extrafin, grå, L×B: 115×150 mm
UOM: 1 * 4 Rulle
Artikel nummer : (MMMASB9000)
Leverantör: 3M
Beskrivning: The Scotch-Brite™ All-purpose scouring pad 9000 is a thick, hand-sized scouring pad designed for cleaning most types of surfaces without scratching.
UOM: 1 * 40 Items
Leverantör: Sampling Systems
Beskrivning: These cleaning validation coupons are available in all popular materials and various sizes.
Leverantör: Sampling Systems
Beskrivning: Cleaning coupons are great for validating a cleaning regime.
Artikel nummer : (MMMA7100262369)
Leverantör: 3M
Beskrivning: Scotch-Brite™ Diamond floor brush 1000 series is designed to clean and polish uncoated concrete, terrazzo and potentially other stone floors in one st...
UOM: 1 * 2 Items
New Product
Artikel nummer : (MMMA2000HEX)
Leverantör: 3M
Beskrivning: The Scotch-Brite™ Low-scratch scouring pad 2000HEX is a powerful two-in-one scouring pad.
UOM: 1 * 15 Items
Artikel nummer : (MMMAHP350)
Leverantör: 3M
Beskrivning: Scotch-Brite™ Extra-thick delicate surface cleaning pads 350 are made from a high-quality synthetic fibre with a non-woven open structure. Non-abrasiv...
UOM: 1 * 40 Items
Artikel nummer : (MMMA7100262371)
Leverantör: 3M
Beskrivning: Scotch-Brite™ Diamond floor brush 3000 series is designed to clean and polish uncoated concrete, terrazzo and potentially other stone floors in one st...
UOM: 1 * 2 Items
New Product
Artikel nummer : (MMMA07440)
Leverantör: 3M
Beskrivning: Rengöringsdynor, Scotch-Brite™, Brun, L×B: 224×158 mm
UOM: 1 * 20 Items
Artikel nummer : (MMMA63119)
Leverantör: 3M
Beskrivning: These abrasive sheets can be used for the intermediate and final sanding of furniture parts, varnished surfaces, kitchen counter-tops and plastic part...
UOM: 1 * 50 Items
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 25
no targeter for Bottom