Vill du logga in igen?
Desiccator cabinets, vertical, secador®
Leverantör: Bel-Art
Secador®
Dessicator cabinets which can be used with disposable or reusable Scienceware® desiccant for safe storage of moisture sensitive products. Desiccant can be placed on or under the perforated bottom shelf. The door opening is 11,4×22,5 cm and includes latch and 2 loops for seals.
Clear
Portable cabinets have convenient built in carrying handles
Vertical cabinets are interchangably stackable
Door can be secured with tamper evident tags or padlock
Easy to view dial hygrometer in the door
ORDER
Expandera alla / Komprimera alla
Benämning Antal hyllor B×D×H, utv Vikt Förp. VWR art.nr Antal
Vertical desiccator, clear, 21 l 1(fixed) 341×410×213 mm 7.2 kg 1 BELAF420710000
Vertikal exsickator, genomskinlig, med blå ändpaneler, 21 l 1(fixed) 341×410×213 mm 7.2 kg 1 BELAF420710006
Vertikal exsickator, brun, 21 l 1(fixed) 341×414×213 mm 7.2 kg 1 BELAF420710008
Vertikal exsickator, genomskinlig, 32,8 l 2 341×414×314 mm - 1 BELAF420721000
Vertikal exsickator, genomskinlig, 43,8 l 3 341×414×417 mm - 1 BELAF420731000
Vertikal exsickator, brun, 43,8 l 3 341×414×417 mm - 1 BELAF420731008
Vertikal exsickator, genomskinlig, 53 l 3 341×414×518 mm - 1 BELAF420741000
Vertikal exsickator, genomskinlig, med blå ändpaneler, 53 l 3 341×414×518 mm - 1 BELAF420741006
Vertikal exsickator, blå, med genomskinlig dörr, 53 l 3 341×414×518 mm - 1 BELAF420741007
Vertikal exsickator, brun, 53 l 3 341×414×518 mm - 1 BELAF420741008
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal