Vill du logga in igen?
Powder funnels
Leverantör: Bel-Art
SCIENCEWARE®
PP, translucent.
PP, translucent
Specially designed for transferring powders. These funnels have 60º angle and a wide stem for clog-free filling.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
Specially designed for transferring powders. These funnels have 60º angle and a wide stem for clog-free filling.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
ORDER
Expandera alla / Komprimera alla
Tratt, y.d. Rör, y.d. Rörlängd Förp. VWR art.nr Antal
65 mm 15 mm 25 mm 12 BELAH146600065
80 mm 18 mm 25 mm 12 BELAH146600080
100 mm 21 mm 26 mm 6 BELAH146600100
142 mm 29 mm 30 mm 4 BELAH146600150
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal