Du sökte efter: Färger för Elektrofores
55  results were found
SearchResultCount:"55"
Sort Results
Visa lista Easy View (ny)
Ge feedback på sökresultat
Leverantör: VWR Chemicals
Beskrivning: These DNA loading dyes are used to load DNA samples to agarose or SDS DNA gels for gel electrophoresis.
Artikel nummer : (1B1680-0.5ML)
Leverantör: VWR Chemicals
Beskrivning: EZ-Vision BlueLight is a sensitive, non mutagenic and environmentally safe fluorescent dye designed for gel staining.
UOM: 1 * 0,5 mL
Leverantör: VWR Chemicals
Beskrivning: VividPro Plus™ is a fluorescent protein stain that can be used in a fast co-electrophoretic protocol to visualize proteins in SDS PAGE gels.
Artikel nummer : (1B1812-25ML)
Leverantör: VWR Chemicals
Beskrivning: This loading dye base is used for custom loading buffer preparation. It is a concentrated solution containing patent blue, bromophenol blue and amaran...
UOM: 1 * 25 mL
Artikel nummer : (N734-5ML)
Leverantör: VWR Chemicals
Beskrivning: Gold-N-Gel RNA dye is compatible with standard protocols for denaturing electrophoresis of RNA fragments. Bands are immediately visible upon illuminat...
UOM: 1 * 5 mL
Safer Human and Environmental Health
Leverantör: VWR Chemicals
Beskrivning: VividPro™ is a generic fluorescent stain for protein gel electrophoresis that can be used to visualize proteins.
Artikel nummer : (E190-5ML)
Leverantör: VWR Chemicals
Beskrivning: Designed to optimise loading nucleic acids in agarose gels, the loading dye imparts colour to the sample to facilitate the loading process and increas...
UOM: 1 * 5 mL
Leverantör: VWR Chemicals
Beskrivning: EZ-Vision® DNA dye as loading buffer provides a non mutagenic, environmentally friendly alternative to ethidium bromide for instantaneous fluorescent ...
Artikel nummer : (K217-1L)
Leverantör: VWR Chemicals
Beskrivning: Blue BANDit is a safe and environmentally friendly alternative to traditional Coomassie® Blue staining.
UOM: 1 * 1 L
Leverantör: Biotium
Beskrivning: GelRed and GelGreen are highly sensitive DNA/RNA gel stains made safe by preventing penetration into cells.
Leverantör: G-Biosciences
Beskrivning: A single step stain for the rapid detection of proteins resolved by PAGE. The stain is based on the interaction of zinc ions with polyacrylamide and p...
Artikel nummer : (786-254)
Leverantör: G-Biosciences
Beskrivning: The stain uses an enhanced Periodic Acid-Schiff (PAS) method for detection of glycoprotein sugars. The supplied agent first oxidises the cis-diol sug...
UOM: 1 * 1 KIT
Artikel nummer : (R0631)
Leverantör: Thermo Fisher Scientific
Beskrivning: Oange loading dye (6X) is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. Ideal for conventional DNA electropho...
UOM: 1 * 1 KIT
Leverantör: Biotium
Beskrivning: Stains specifically designed for DNA in polyacrylamide gels.
Leverantör: G-Biosciences
Beskrivning: LabSafe GEL Blue™ is an enhanced protein stain based on Coomassie dye that offers unsurpassed sensitivity and rapid band visualisation.
Artikel nummer : (R0611)
Leverantör: Thermo Fisher Scientific
Beskrivning: This gel loading dye contains two different dyes bromophenol blue and xylene cyanol FF for visual tracking of DNA migration during electrophoresis. Th...
UOM: 1 * 1 KIT
Fråga efter pris
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
Lager för denna produkt är begränsat, men kan vara tillgänglig i ett lager nära dig. Se till att du är inloggad på hemsidan så att tillgängligt lager kan visas. Om call fortfarande visas och du behöver hjälp, ring oss på 08-621 34 20.
VWR International AB säljer bara etanol till skattebefriade kunder. Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via e-post.
VWR International AB säljer bara etanol till skattebefriade kunder.
Denna produkt ä reglerad enligt svensk lagstiftning. Om ytterligare information krävs av dig kommer du att kontaktas via email.
Produkten har blockerats av din organisation. Vänligen kontakta din inköpsavdelning för mer information.
Lösenordet och säkerhetsfrågan associerade med ditt Kundnummer har gått ut. Vänligen ändra ditt lösenord och din säkerhetsfråga.
Denna produkt är inte längre tillgänglig. Alternativ kan vara tillgängliga genom att söka med VWR art.nr ovan. Om du behöver ytterligare hjälp, ring VWR Kundservice på 08-621 34 20.
1 - 16 of 55
no targeter for Bottom