Tanks, cylindrical, with cover
Leverantör: Bel-Art
BELAH351100000EA 4050 SEK
BELAH351100000
Tanks, cylindrical, with cover
This heavy-duty, molded PE plastic tanks helps to safely hold chemicals and hot liquids up to 52 °C.
  • Resistant to rust, corrosion and dents
  • Capacity marked on both the tank and the lid
  • Lids have a central grip knob for easy removal and replacement
Open-top tanks are stress-free and self-supporting with a seamless wall about 4,8 mm thick.
Certifikat: Meets FDA requirements for food handling.
Beställningsinformation: Polly-Crocks with a sight glass can be ordered separately.