First aid panel, small
Leverantör: Plum Safety
118-0227EA 880 SEK
PLUM5101
First aid panel, small
All in one solution for virtually any workspace. Optimal care for eye injuries and cuts.
Contents:
  • Eye wash bottle, 500 ml, sterile and maintenance-free (0,9% sodium chloride solution), with ergonomic eye cup and wall bracket
  • QuickFix plaster dispenser, including 45 elastic and 45 waterproof PE plasters
  • Quick Clean wound cleansing wipes, 40 pcs in dispenser box, individually wrapped
  • With wall bracket