Vill du logga in igen?
Precision alcohol meters
Leverantör: ALLA ATELIERS
These high precision alcoholometers with individual number on the scale and a calibration temperature of 20 °C.
Certifikat: ISO 4801 and NF B 35-515
ORDER
Expandera alla / Komprimera alla
Mätområde Indelning Längd Förp. VWR art.nr Antal
0 - 10% EtOH 0,1 % 350 mm 1 614-0004
5 - 15% EtOH 0,1 % 350 mm 1 ALLA0910FC005/20QP
6 - 16% EtOH 0,1 % 350 mm 1 ALLA0910FC006/20QP
10 - 20% EtOH 0,1 % 350 mm 1 614-0006
20 - 30% EtOH 0,1 % 350 mm 1 614-0007
30 - 40% EtOH 0,1 % 350 mm 1 ALLA0910FC030/20QP
40 - 50% EtOH 0,1 % 350 mm 1 ALLA0910FC040/20QP
45 - 55% EtOH 0,1 % 350 mm 1 ALLA0910FC045/20QP
50 - 60% EtOH 0,1 % 350 mm 1 614-0013
50 - 65% EtOH 0,1 % 350 mm 1 ALLA0910FC055/20QP
60 - 70% EtOH 0,1 % 350 mm 1 614-0015
65 - 75% EtOH 0,1 % 350 mm 1 ALLA0910FC065/20QP
70 - 80% EtOH 0,1 % 350 mm 1 614-0017
75 - 85% EtOH 0,1 % 350 mm 1 ALLA0910FC075/20QP
80 - 90% EtOH 0,1 % 350 mm 1 614-0021
85 - 95% EtOH 0,1 % 350 mm 1 ALLA0910FC085/20QP
90 - 100% EtOH 0,1 % 350 mm 1 614-0022
95 - 103% EtOH 0,1 % 350 mm 1 ALLA0910FC095/20QP
0 - 20% EtOH 0,2 % 350 mm 1 ALLA0920FC000/20QP
20 - 40% EtOH 0,2 % 350 mm 1 ALLA0920FC020/20QP
40 - 60% EtOH 0,2 % 350 mm 1 ALLA0920FC040/20QP
60 - 80% EtOH 0,2 % 350 mm 1 ALLA0920FC060/20QP
80 - 100% EtOH 0,2 % 350 mm 1 ALLA0920FC080/20QP
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal