Vill du logga in igen?
Retort rods, with M10 thread
Leverantör: NICKEL ELECTRO
Aluminium, mild steel or stainless steel.
ORDER
Expandera alla / Komprimera alla
Diam. Längd Material Gänga Förp. VWR art.nr Antal
10 mm 500 mm Rostfritt stål M10 1 NICK8624
10 mm 500 mm Stål M10 1 NICK8634
12,5 mm 600 mm Stål M10 1 NICK8631
12,5 mm 750 mm Stål M10 1 NICK8632
12,5 mm 1000 mm Stål M10 1 NICK8633
12 mm 500 mm Aluminium M10 1 NICK8614
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal