Methyl methacrylate-[D₈] (monomer) (99.5% D) stabilised for NMR spectroscopy
Leverantör: Merck
|
Fara
|
8.15050.0005EA
0
SEK
8.15050.0005
Methyl methacrylate-[D₈] (monomer) (99.5% D) stabilised for NMR spectroscopy
Methyl methacrylate-[D₈] (monomer)
Formel:
C₅D₈O₂ |
CAS nummer:
35233-69-3 EINECS: 252-449-9 UN: 1247 ADR: 3,II |