Vill du logga in igen?
Gas cartridge filters, Super Clean™
Leverantör: Thermo Scientific
Super Clean™
These filters can be easily replaced without tools.
  • 99,9999% pure gas (or 6,0 grade) output
  • No carrier gas line contamination during cartridge change
  • Maximum pressure of 11 bar (160 psi)
  • Maximum flow of 7 l/min
Certifikat: TUEV approved.
ORDER
Expandera alla / Komprimera alla
Cartridge filter baseplates/manifolds
Benämning Förp. För VWR art.nr Antal
Cartridge filter base plate, single base 1 Carrier gas only, for triple filter HYPE60180-814
Cartridge filter base plate, dual base 1 Air and fuel gas, for 2 × combi filter HYPE60180-815
Cartridge filter base plate, triple base 1 Carrier, make-up and fuel gas, for triple and 2 × combi filter HYPE60180-817
Cartridge filter base plate, two-position, high-flow base 1 Carrier, make-up and fuel, for N₂ in LC/MS, H2O in H2/air generator HYPE60180-816
Cartridge filter base plate, four-position base 1 High capacity carrier, fuel, make-up, for moisture, oxygen, 2 × hydrocarbon HYPE60180-818
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
TILLBEHÖR
Expandera alla / Komprimera alla
Accessories
Benämning Förp. VWR art.nr Antal
O-ring 20 HYPE60180-837
Flush cap 1 HYPE60180-838
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indikera Batch Nummer. Detta fält är obligatoriskt. Ange "x" om detta inte finns för VWR kemikalier
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Eller
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beskrivning Artikel nummer Availability Enhet Ditt pris Price Per Antal
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping